Anti-CA 19-9 Mouse Monoclonal Antibody [clone: 13H12]
Lieferant: US Biological
137580
USBI137580EA 688 EUR
USBI137580
Anti-CA 19-9 Mouse Monoclonal Antibody [clone: 13H12]
Antikörper
Type: Primary
Antigen: CA 19-9
Clonality: monoclonal
Clone: 13H12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Bestellen