Anti-IBSP Rabbit Polyclonal Antibody

Lieferant: US Biological
144247
USBI144247EA 587 EUR
USBI144247
Anti-IBSP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IBSP (integrin-binding sialoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR