Anti-IDH1 Rabbit Polyclonal Antibody

Lieferant: US Biological
146682
USBI146682EA 574 EUR
USBI146682
Anti-IDH1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-2?g/ml

Type: Primary
Antigen: IDH1 (isocitrate dehydrogenase 1 (NADP+), soluble)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR