Maackia amurensis Lectin (MAA/MAL II) (FITC)
Lieferant: US Biological
518542
USBI518542EA 446 EUR
USBI518542
Maackia amurensis Lectin (MAA/MAL II) (FITC)
Proteinkonjugate und Markierungen
Maackia amurensis Lectin (MAA/MAL II) is labelled with fluorescein isothiocyanate (FITC) and has an appropriate number of fluorochromes bound to provide the optimum staining characteristics for this lectin.
  • Source: Amur maackia
  • Inhibitory carbohydrate: Human Glycophorin
  • Carbohydrate specificity: Siaα2-3Galβ1-3GalNAc
FITC conjugates have been used in a variety of immunohistochemical and flow cytometry applications. Fluorescein conjugates display a high rate of photobleaching, signal is sensitive to pH changes, broad fluorescence emission spectrum, and fluorescence quenching on conjugation to biopolymers.
Store product at 4 °C if to be used immediately within two weeks. For long-term storage, aliquot to avoid repeated freezing and thawing and store at –20 °C. Aliquots are stable at –20 °C for 6 months after receipt. Dilute required amount only prior to immediate use. Further dilutions can be made in assay buffer. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.
Bitte beachten FITC conjugates are sensitive to light. This product as supplied is intended for research use only, not for use in human, therapeutic or diagnostic applications.