Anti-AIF1 Goat Polyclonal Antibody

Lieferant: US Biological
A1059-86G
USBIA1059-86GEA 601 EUR
USBIA1059-86G
Anti-AIF1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 0.5-1. 5 ?g/ml; Immunochemistry: 2-4?g/ml

Type: Primary
Antigen: AIF1 (allograft inflammatory factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR