Anti-IAPP Rabbit Polyclonal Antibody
Lieferant: US Biological
A2275-52J
USBIA2275-52JEA 540 EUR
USBIA2275-52J
Anti-IAPP Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 1:200; Immunochemistry: 1:500
Type: Primary
Antigen: IAPP (islet amyloid polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Cat, Mouse, Rat