Anti-IAPP Rabbit Polyclonal Antibody
Lieferant: US Biological
A2275-52M
USBIA2275-52MEA 883 EUR
USBIA2275-52M
Anti-IAPP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IAPP (islet amyloid polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat