Anti-IAPP Rabbit Polyclonal Antibody

Lieferant: US Biological
A2275-52N
USBIA2275-52NEA 837 EUR
USBIA2275-52N
Anti-IAPP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IAPP (islet amyloid polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR