Anti-APP Mouse monoclonal antibody unconjugated [clone: 13A237]

Lieferant: US Biological
A2275-86S
USBIA2275-86SEA 485 EUR
USBIA2275-86S
Anti-APP Mouse monoclonal antibody unconjugated [clone: 13A237]
Antikörper
Recommended dilution: Immunochemistry: 40-150 ?g

Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: 13A237
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR