Anti-Catenin, delta 1 Rabbit Polyclonal Antibody

Lieferant: US Biological
C2069-54M
USBIC2069-54MEA 675 EUR
USBIC2069-54M
Anti-Catenin, delta 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Catenin, delta 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR