Anti-ITGAX Hamster Monoclonal Antibody

Lieferant: US Biological
C2262-57F-500
USBIC2262-57F-500EA 499 EUR
USBIC2262-57F-500
Anti-ITGAX Hamster Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ITGAX (integrin, alpha X (complement component 3 receptor 4 subunit))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR