Anti-ITGAE (Integrin alpha E, CD103) Hamster Monoclonal Antibody (Biotin) [clone: 2E7]

Lieferant: US Biological
C2445-65E
USBIC2445-65EEA 513 EUR
USBIC2445-65E
Anti-ITGAE (Integrin alpha E, CD103) Hamster Monoclonal Antibody (Biotin) [clone: 2E7]
Antikörper
Type: Primary
Antigen: ITGAE (Integrin alpha E, CD103)
Clonality: Monoclonal
Clone: 2E7
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR