Anti-Death-associated protein kinase Rabbit polyclonal antibody

Lieferant: US Biological
D1051-02
USBID1051-02EA 608 EUR
USBID1051-02
Anti-Death-associated protein kinase Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Death-associated protein kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR