Anti-Dapsone Sheep polyclonal antibody
Lieferant: US Biological
D1065-01
USBID1065-01EA 763 EUR
USBID1065-01
Anti-Dapsone Sheep polyclonal antibody
Antikörper
Recommended dilution: ELISA: 24?g/ml
Type: Primary
Antigen: Dapsone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Bestellen