Anti-Defender of Apoptotic Cell Death 1 Rabbit polyclonal antibody

Lieferant: US Biological
D1877-15B
USBID1877-15BEA 716 EUR
USBID1877-15B
Anti-Defender of Apoptotic Cell Death 1 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 0.5-2.0 ?g/ml

Type: Primary
Antigen: Defender of Apoptotic Cell Death 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR