Drop-out mix synthetic minus adenine, histidine, leucine, tryptophan, uracil without yeast nitrogen base
Lieferant: US Biological
D9540-05
USBID9540-05EA
203
EUR
USBID9540-05
Drop-out mix synthetic minus adenine, histidine, leucine, tryptophan, uracil without yeast nitrogen base
Medien für die Mikrobiologie
Additive und Supplemente für die Mikrobiologie
Drop-out mixes are used as a supplement to Drop-out base for making synthetic, complete media formulations in accordance with methods in yeast genetics.
- Convenient powders
- Useful for one-hybrid and two-hybrid systems
All lots are quality control tested using the appropriate yeast strains and transformants.