Anti-FKLF Rabbit polyclonal antibody

Lieferant: US Biological
F4524-02
USBIF4524-02EA 466 EUR
USBIF4524-02
Anti-FKLF Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: FKLF
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR