5-Fluororotsäure Monohydrat

Lieferant: US Biological

Synonyme: 5-FOA

F5050-500
USBIF5050-500EA 87.7 EUR
USBIF5050-500
5-Fluororotsäure Monohydrat
5-Fluororotsäure Monohydrat
Formel: C₅H₃FN₂O₄·1H₂O
Molecular Weight: 174,0 g/mol (anhy)
MDL: MFCD00150658
CAS-Nummer: 220141-70-8
EINECS: 211-876-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR