Anti-GLI2 Rabbit Polyclonal Antibody

Lieferant: US Biological
G2035-53D
USBIG2035-53DEA 812 EUR
USBIG2035-53D
Anti-GLI2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: GLI2 (GLI family zinc finger 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR