Anti-IBSP Goat Polyclonal Antibody

Lieferant: US Biological
I0468-50
USBII0468-50EA 871 EUR
USBII0468-50
Anti-IBSP Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.5-1 ?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: IBSP (integrin-binding sialoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR