Anti-IBSP Rabbit Polyclonal Antibody
Lieferant: US Biological
I0468-50A
USBII0468-50AEA 608 EUR
USBII0468-50A
Anti-IBSP Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50
Type: Primary
Antigen: IBSP (integrin-binding sialoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat