Anti-KLF2 Mouse Monoclonal Antibody [clone: 11n121]

Lieferant: US Biological
K1891-37C
USBIK1891-37CEA 817 EUR
USBIK1891-37C
Anti-KLF2 Mouse Monoclonal Antibody [clone: 11n121]
Antikörper
Recommended dilution: Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: KLF2 (Kruppel-like factor 2)
Clonality: monoclonal
Clone: 11n121
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR