Anti-MHC Class 2, I-Ak/s Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F125]
Lieferant: US Biological
M3887-54G
USBIM3887-54GEA 609 EUR
USBIM3887-54G
Anti-MHC Class 2, I-Ak/s Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10F125]
Antikörper
Recommended dilution: Flow Cytometry: 1:5;
Type: Primary
Antigen: MHC Class 2, I-Ak/s
Clonality: monoclonal
Clone: 10F125
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat