Anti-MHC Class 2, I-Ak/s Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10F125]

Lieferant: US Biological
M3887-54H
USBIM3887-54HEA 599 EUR
USBIM3887-54H
Anti-MHC Class 2, I-Ak/s Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10F125]
Antikörper
Recommended dilution: Flow Cytometry:1:10;
Do not freeze!

Type: Primary
Antigen: MHC Class 2, I-Ak/s
Clonality: monoclonal
Clone: 10F125
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR