Anti-MSRA Rabbit Polyclonal Antibody

Lieferant: US Biological
M4692-59D
USBIM4692-59DEA 649 EUR
USBIM4692-59D
Anti-MSRA Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: MSRA (methionine sulfoxide reductase A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR