Anti-NOTCH4 Hamster Monoclonal Antibody [clone: 12G40]

Lieferant: US Biological
N5375-51H
USBIN5375-51HEA 367 EUR
USBIN5375-51H
Anti-NOTCH4 Hamster Monoclonal Antibody [clone: 12G40]
Antikörper
Type: Primary
Antigen: NOTCH4 (notch 4)
Clonality: monoclonal
Clone: 12G40
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR