Anti-Parvovirus B19, Human Mouse Monoclonal Antibody [clone: 2Q2041]

Lieferant: US Biological
P3113-81C
USBIP3113-81CEA 485 EUR
USBIP3113-81C
Anti-Parvovirus B19, Human Mouse Monoclonal Antibody [clone: 2Q2041]
Antikörper
Recommended dilution: ELISA: 1:200; Immunofluorescence: 1:200-1:400; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: Parvovirus B19, Human
Clonality: Monoclonal
Clone: 2Q2041
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR