Peroxidase, Horseradish, RZ3 (POD, HRP, HRPO)
Lieferant: US Biological
P3350-5KU
P3350-25KU
P3350-125KU
P3350-10KU
USBIP3350-5KUEA
101
EUR
USBIP3350-5KU
USBIP3350-25KU
USBIP3350-125KU
USBIP3350-10KU
Peroxidase, Horseradish, RZ3 (POD, HRP, HRPO)
Enzyme
Horseradish Peroxidase (HRP) is a hemoprotein catalysing the oxidation by hydrogen peroxide of a number of substrates such as ascorbate, ferrocyanide, cytochrome C and the leuko form of many dyes.
Horseradish Peroxidase has been found well suited for the preparation of enzyme conjugated antibodies, due in part to its ability to yield chromogenic products and to its relatively good stability characteristics.