Anti-Phospholipase A2, Secretory, Group IB Mouse Monoclonal Antibody [clone: 11n151]

Lieferant: US Biological
P4074-06D5
USBIP4074-06D5EA 802 EUR
USBIP4074-06D5
Anti-Phospholipase A2, Secretory, Group IB Mouse Monoclonal Antibody [clone: 11n151]
Antikörper
Recommended dilution: Western Blot: 2?g/ml; Immunoprecipitation: 25 ?g/ml

Type: Primary
Antigen: Phospholipase A2, Secretory, Group IB
Clonality: monoclonal
Clone: 11n151
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR