Anti-TIMP1 Rabbit Polyclonal Antibody

Lieferant: US Biological
T5580-05J
USBIT5580-05JEA 559 EUR
USBIT5580-05J
Anti-TIMP1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 2-4?g/ml

Type: Primary
Antigen: TIMP1 (TIMP metallopeptidase inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR