Human Recombinant tPA (From CHO cells)
Lieferant: US Biological
T5600-07B
USBIT5600-07BEA 776 EUR
USBIT5600-07B
Human Recombinant tPA (From CHO cells)
Proteine und Peptide
This recombinant human tPA is active and expressed in Chinese hamster ovary (Cho) cells. Fully complexable with PAI-1.
  • Activity: >90% active as determined by titration with PAI-1
Bitte beachten Intended for research use only. Not for use in human, therapeutic, or diagnostic applications.