Anti-Tumor Necrosis Factor Receptor 1-Associated Death Domain Protein Isoform 2 Goat Polyclonal Antibody

Lieferant: US Biological
T8170-50D
USBIT8170-50DEA 643 EUR
USBIT8170-50D
Anti-Tumor Necrosis Factor Receptor 1-Associated Death Domain Protein Isoform 2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:128000; Western Blot: 1-3?g/ml

Type: Primary
Antigen: Tumor Necrosis Factor Receptor 1-Associated Death Domain Protein Isoform 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR