Anti-WRN Mouse Monoclonal Antibody [clone: 11C960]

Lieferant: US Biological
W9000-03
USBIW9000-03EA 623 EUR
USBIW9000-03
Anti-WRN Mouse Monoclonal Antibody [clone: 11C960]
Antikörper
Recommended dilution: ELISA: 1 ?g/ml-3ng/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: WRN (Werner syndrome, RecQ helicase-like)
Clonality: monoclonal
Clone: 11C960
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR