Polyethylene Adhesion Tape
Lieferant: ULTRATAPE
1110RE200-P3D
UTAP1110RE200-P3DEA 49.3 EUR
UTAP1110RE200-P3D
Polyethylene Adhesion Tape
Klebebänder
UltraTape 1110 is designed specifically for sealing wafer shipping boxes as well as cleanroom packaging and bag sealing. Its specially formulated medium-high adhesive leaves no residue or ghosting on plastic boxes, even after prolonged storage.