Banded Earplugs, X-fold
Lieferant: UVEX
2125.344
UVEX2125.344EA 32 EUR
UVEX2125.344
Banded Earplugs, X-fold
Gehörschutzstöpsel Bügelgehörschützer
The innovative design with unique, oval-shaped soft foam earplugs guarantees a comfortable fit.
  • X-fold is a lightweight banded ear protector weighing just 8 g
  • SNR 23 dB H: 26 dB, M: 18 dB, L: 17 dB
  • Colour: grey, sky blue, lime
  • Compact fold up, easy to carry
  • Suspended under the chin, eliminating friction sounds resulting from clothing
  • Band made up of polypropylene; earplugs made up of PU-foam
  • Thermal impac: Cold, heat
The foldable band is ergonomically shaped and prevents annoying noises from being transmitted by keeping the band away from the wearers clothes.