Safety spectacles, Polarvision
Lieferant: UVEX
9231.960
UVEX9231.960EA 94.6 EUR
UVEX9231.960
Safety spectacles, Polarvision
Schutzbrillen
Comfortable safety spectacles with polarisation filter.
  • Polarisation filter protects against reflections and glare (e.g. water surfaces and highly polished materials)
  • Scratch-resistant coated
Suitable for workstations with reflective surfaces (offshore, maritime, drivers, construction industry).