Goggles, pheos cx2 sonic
Lieferant: UVEX
9309.275
UVEX9309.275EA 26.6 EUR
UVEX9309.275
Goggles, pheos cx2 sonic
Vollsichtbrillen
With its compact, lightweight design, the uvex pheos cx2 sonic is a welcome companion for any job.
  • Comfortable protection thanks to soft forehead and nose piece moulded directly onto the lens
  • Scratch-proof exterior, permanent anti-fog interior (uvex supravision extreme)
  • Can be easily combined with other PPE
  • Textile headband
These functional wide-vision goggles offer impressive comfort and, thanks to uvex supravision coating technology, provide a clear field of vision even in extreme climates.
Standards: EN 166; EN 170