Schuhdesinfektionsspraye, uvex nano shoe add in
Lieferant: UVEX
9698.300
UVEX9698.300EA 22 EUR
UVEX9698.300
Schuhdesinfektionsspraye, uvex nano shoe add in
Schuhcreme und -bürsten
This shoe disinfectant formula contains active ingredients which are approved for use for a number of years and can be found in cosmetics (formaldehyde-free, phenol-free). Unlike deodorants, which simply cover sweaty smells, this product deals with the underlying causes: bacteria.
  • Effective against odour and athelete's foot
  • Contains antimicrobial and antifungal active ingredients which eliminate bacteria and fungus over the longer term
  • Prevents existing fungi and its reoccurrence
For use on the inside surfaces of all footwear.