Viewing cabinets, Chromato-Vue®, C-70G
Lieferant: UVP ULTRA VIOLET PRODUCTS
95-0020-05
UVPA95-0020-05EA 2420 EUR
UVPA95-0020-05
Viewing cabinets, Chromato-Vue®, C-70G
Beleuchtung UV-Betrachtungsgeräte
The large, lightweight plastic UV Chromato-Vue® viewing cabinet C-70G is designed for examining fluorescent samples in a fully lit environment.
  • Cabinets include two 15-watt shortwave (254 nm) and two 15-watt longwave (365 nm) high intensity UV tubes mounted on one side of the cabinet
  • Soft rubber viewport includes a Contrast Control Filter, which absorbs UV for viewing samples fatigue-free with no 'blue haze'
  • Removable bottom panel so the cabinet may be placed over large samples or a transilluminator
  • Access opening via a lever-lift curtain which also blocks ambient light
  • Control switches are conveniently located for selecting either of the UV wavelengths or the interior overhead white light
W×D×H (int.): 432×356×127 mm; W×D×H (ext.): 594×400×330 mm
Weight: 10,3 kg