Stable ozone generators
Lieferant: UVP ULTRA VIOLET PRODUCTS
97-0066-02 97-0067-02
UVPA97-0066-02EA 1060 EUR
UVPA97-0066-02 UVPA97-0067-02
Stable ozone generators
Gasgeneratoren Ozon-Generatoren
Ozone generators produce a continuous flow of ozone as well as a stable source for water purification and air pollution studies. Designed for calibration of field and laboratory ozone analysers in water purification and air pollution applications.
The stable ozone generator utilises the photochemical reaction of oxygen under shortwave UV to produce a continuous flow of ozone. Generators are a stable source for instrument development and an oxidant source for aging and oxidant testing of materials. Models SOG-1 and SOG-2 incorporate Pen-Ray® lamps, which are inserted into the generator boxMore Product Information