IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥98% (durch HPLC auf Trockenbasis), Biotechreagenz, cGMP Herstellung, J.T.Baker®

Lieferant: Avantor
Achtung

V264-05EA 80040 EUR
V264-05 V264-02 V264-01
IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥98% (durch HPLC auf Trockenbasis), Biotechreagenz, cGMP Herstellung, J.T.Baker®
Formel: C₉H₁₈O₅S
Schmelzpunkt: 110…122 °C
MDL: MFCD00063273
CAS-Nummer: 367-93-1

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product Passes Test
Store and ship product between 2 and 8 °C 5.00 - 7.00
CAUTION: For Manufacturing, processing or repackaging ≤1%
Appearance (white to off-white solid) Passes Test
Identification (IR) Passes Test
pH of 5% Solution 5.00 - 7.00
Specific Rotation @ 25 °C -34.5 - -28.5 Degree
Melting Point 110 - 122 °C
Water (H₂O), For Information Only ≤5000 ppm
Purity (by HPLC) (anhydrous basis) ≥98 %
Assay (by HPLC) (anhydrous basis) 98 - 102
Endotoxin Concentration, EU/g –34.5 - –28.5°
Residual Methanol ≤3000 ppm
Residual Ethanol ≤5000 ppm
Residual Isopropanol ≤5000 ppm
Residual Ethyl Acetate ≤5000 ppm
Total Aerobic Microbial Count ≤100 cfu/g
Total Yeast and Mold Count ≤100 cfu/g
Biological Induction Characterization (induction response @ 1mM)
Arsenic (As), For Information Only
Cadmium (Cd), For Information Only
Mercury (Hg), For Information Only
Lead (Pb), For Information Only
Cobalt (Co), For Information Only
Nickel (Ni), For Information Only
Vanadium (V), For Information Only
Silver (Ag), For Information Only
Gold (Au), For Information Only
Iridium (Ir), For Information Only
Osmium (Os), For Information Only
Palladium (Pd), For Information Only
Platinum (Pt), For Information Only
Rhodium (Rh), For Information Only
Ruthenium (Ru), For Information Only
Selenium (Se), For Information Only
Thallium (Tl), For Information Only
Barium (Ba), For Information Only
Chromium (Cr), For Information Only
Copper (Cu), For Information Only
Lithium (Li), For Information Only
Molybdenum (Mo), For Information Only
Antimony (Sb), For Information Only
Tin (Sn), For Information Only
Aluminum (Al), For Information Only
Iron (Fe), For Information Only
Manganese (Mn), For Information Only
Zinc (Zn), For Information Only
Residual Solvents: The Class 3 solvents Ethanol, Isopropanol, Ethyl
Acetate and the Class 2 solvent Methanol are likely to be present. These solvents are quantified and reported on each batch. No other
Class 1,2,3, or other solvents are used or produced in the production
process. Dioxane was not used in the manufacturing process.
Bacterial Induction characterization: Test performed at 1 mM of IPTG (E.
Coli. protein expression)
IPTG is highly hygroscopic. Allow packaging/container to equilibrate to
room temperature before opening for analysis. Ensure sample is kept dry
(such as in a desiccator) after opening.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR