Tetrahydrofuran 99,8, UltimAR® LC, UV, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Tetramethylenoxid, Furantetrahydrid, 1,4-Oxidobutan, Oxolan, THF

V558-10EA 0 EUR
V558-10
Tetrahydrofuran 99,8, UltimAR® LC, UV, Macron Fine Chemicals™
Tetrahydrofuran
Formel: C₄H₈O
Molecular Weight: 72.11 g/mol
Siedepunkt: 66
Schmelzpunkt: –108.5 °C
Dichte: 0.888 g/cm³ (20 °C)
Flammpunkt: –21.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005356
CAS-Nummer: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Assay (C₄H₈O) (by GC, corrected for water) ≥99.8 %
Color (APHA) ≤10
Peroxide (as H₂O₂)(at time of packaging) ≤0.005 %
Residue after Evaporation ≤2.0 ppm
Water (H₂O)(by KF, coulometric) ≤0.02 %
Optical Absorbance (1-cm path vs water) - 212 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 230 nm ≤0.40
Optical Absorbance (1-cm path vs water) - 254 nm ≤0.15
Optical Absorbance (1-cm path vs water) - 280 nm ≤0.03
Optical Absorbance (1-cm path vs water) - 320 - 400 nm ≤0.005
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR