Centring and sealing rings for small flange connections
Lieferant: VACUUBRAND
20660140 20660141 20660143
VACU20660140EA 13.5 EUR
VACU20660140 VACU20660141 VACU20660143
Centring and sealing rings for small flange connections
Versiegeln Dichtringe
These centring and sealing rings for small stainless steel flange KF connections. With aluminium sealing ring.