Chemistry diaphragm pumps and chemistry vacuum systems with ATEX approval, VARIO®
Lieferant: VACUUBRAND
20696936 20696937 20696946 20696947
VACU20696936EA 9810 EUR
VACU20696936 VACU20696937 VACU20696946 VACU20696947
Chemistry diaphragm pumps and chemistry vacuum systems with ATEX approval, VARIO®
Pumpen Vakuum-Membranpumpen
For use in explosive areas chemistry diaphragm pumps category 2 with ATEX conformity are available.
  • Full advantage of ATEX chemistry diaphragm pumps
  • VARIO®: short process times due to hysteresis-free VARIO® vacuum control
  • VARIO® pump operates on demand only as fast as necessary – minimal power consumption, extended maintenance intervals, silent operation
  • Noise level: 42 dB(A)
  • Electrical: 230 V, 50 Hz
In explosive areas, only operating materials with ATEX conformity may be used. VACUUBRAND offers chemistry diaphragm pumps and pumping units category 2 (More Product Information