Chemistry vacuum pump, NT series
Lieferant: VACUUBRAND
743301 20743300 20743500 743501 743302 743602 743601 20743600 743502
VACU743301EA 7040 EUR
VACU743301 VACU20743300 VACU20743500 VACU743501 VACU743302 VACU743602 VACU743601 VACU20743600 VACU743502
Chemistry vacuum pump, NT series
Pumpen Vakuum-Membranpumpen
Compact, powerful pump gives ideal solution for continuous, oil-free pumping of corrosive gases and vapors with exceptional high pumping speed. Eight-cylinder NT pump features quiet operation with smooth, easy-to-clean surfaces.
  • High chemical and vapour resistance
  • Very high pumping speed even at low vacuum levels
  • Long diaphragm life with maintenance free drive system
  • Ultimate vacuum even with gas ballast
  • PTFE sandwich diaphragms for increased reliability and extended operating life
  • All internal parts are made of chemically resistant fluoroplastics
  • Noise
More Product Information