Rotary Vane Pumps, RZ 9
Lieferant: VACUUBRAND
20798140
VACU20798140EA 3950 EUR
VACU20798140
Rotary Vane Pumps, RZ 9
Pumpen Vakuum-Drehschieberpumpen
Rotary vane pumps from VACUUBRAND offer great performance in a compact design. Even with the gas ballast open, they enable a very good ultimate vacuum. You also benefit from extended oil change and maintenance intervals thanks to optimized lubrication circuit and large usable oil volume.
  • Very high flow rates even at vacuum levels approaching ultimate vacuum
  • High water vapor tolerance due to efficient gas ballast
  • Very good ultimate vacuum even with gas ballast
  • Vacuum-tight at switch-off
  • External anti-suckback valve not needed
  • Large oil volume: long
More Product Information