Solid phase extraction cartridges, Bond Elut™ Mycotoxin
Lieferant: VARIAN
12165001B
VARI12165001BEA 637 EUR
VARI12165001B
Solid phase extraction cartridges, Bond Elut™ Mycotoxin
Chromatographiesäulen
Bond Elut™ Mycotoxin is a novel sorbent which cleans up food extracts for improved trichothecene and zearalenone analysis. Results are comparable or superior to competing methods, including immunoaffinity columns (IaC) and charcoal/alumina columns. The sorbent is a proprietary silica-based ion exchange material.
  • Simple methodology saves time and increases throughput
  • Use with a broad range of food matrices
  • Alternative to immunoaffinity techniques
The Bond Elut™ Mycotoxin method for extraction and cleanup is successful with a variety of food and grain sample types, including wheat, corn, durum, oats, bread, muesli and infant food.
Bond Elut™ Mycotoxin is easy to use and acts in a selective non-retention way – the toxin analytes pass through the cartridge while the food matrix components are retained.