Tuning and performance standards for LC/MS
Lieferant: VARIAN
393210201
VARI393210201EA 252 EUR
VARI393210201
Tuning and performance standards for LC/MS
Standards Organische Standards
Agilent offers a range of tuning and performance standards for LC/MS.
  • Typical standards have at least a year of shelf-life
  • Many standards align with a range of applications
  • Unique formulations that are tested and verified on Agilent instrumentation
A critical part of developing a solution, keeping your instrument qualified, and even trusting the instrument’s results is to regularly test the efficacy of the instrument. Agilent offers a range of certified chemical standards that are integral to instrument, performance, and application verification for your LC and LC/MS system. You can rest assured that Agilent tuning mixes are of the highest quality –designed, developed, and manufactured to guarantee at least one year of shelf-life.
Lieferumfang: Most standards manufactured with ISO 17025 and Guide 34 certification.
Verpackung: Environmentally friendly packaging that is easy-to-open and verify information.