Cary source lamps
Lieferant: VARIAN
5610013900 5610021700 110713990
VARI5610013900EA 56 EUR
VARI5610013900 VARI5610021700 VARI110713990
Cary source lamps
Beleuchtung Deuterium-Lampen
Source lamps for Cary spectrophotometers are fully modular and interchangeable. A range of UV, xenon and visible lamps are available.
  • Fully modular and compatible with Cary spectrophotometers
The UV, xenon and visible lamps are fully modular, allowing you to select and change between light sources according to your experimental requirements.