Graphite tubes and platforms
Lieferant: VARIAN
6310001200 6310001100 6310001300
636-2501EA 614 EUR
636-2501 VARI6310001100 VARI6310001300
Graphite tubes and platforms
Chromatographie-Zubehör Chromatographie-Ersatzteile
Graphite tubes are quality tested to ensure each batch passes our demanding performance specifications for contamination, sensitivity, precision, electrical resistance and lifetime.
  • High purity graphite reduces spurious absorption from trace contaminants and improves signal-to-noise
  • Every tube is hand-checked for reproducible and reliable results
  • Perform fewer repeats and increase productivity
Use partitioned tubes for general purpose work and when you need the best detection limits with samples that have low background. These are also ideal whenMore Product Information
Bestellen
TECHNISCHE DATEN
Environmentally Preferable